Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stox1
Entrez Gene ID
294388
Gene Name
storkhead box 1
Gene Aliases
RGD1310622
Chromosome Location
Chr.20: 32276857 - 32353677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191720.1 NP_001178649.1 2 - 3 455 85

View Full Product Details