Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phactr1
Entrez Gene ID
306844
Gene Name
phosphatase and actin regulator 1
Gene Aliases
-
Chromosome Location
Chr.17: 23245423 - 23761207 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214457.2 NP_999622.2 10 - 11 1633 69
GenBank mRNA AY494977.1 - - 1502 69
BC098634.1 - 10 - 11 1633 69

View Full Product Details