The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153738.1 | NP_714960.1 | 5 - 6 | 568 | 110 |
GenBank mRNA | AF525159.1 | - | 5 - 6 | 568 | 110 |