Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm24
Entrez Gene ID
690139
Gene Name
RNA binding motif protein 24
Gene Aliases
-
Chromosome Location
Chr.17: 18671037 - 18682876 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191100.1 NP_001178029.1 3 - 4 432 59

View Full Product Details