Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd3
Entrez Gene ID
306810
Gene Name
sushi domain containing 3
Gene Aliases
-
Chromosome Location
Chr.17: 15814132 - 15830328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107341.1 NP_001100811.1 2 - 3 310 77
XM_006253709.2 XP_006253771.1 2 - 3 1886 77
GenBank mRNA FQ227785.1 - 2 - 3 495 77

View Full Product Details