Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Susd3
Entrez Gene ID
306810
Gene Name
sushi domain containing 3
Gene Aliases
-
Chromosome Location
Chr.17: 15814132 - 15830328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107341.1 NP_001100811.1 3 - 4 456 84
XM_006253709.2 XP_006253771.1 3 - 4 2032 84

View Full Product Details