Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
296623
Gene Name
family with sequence similarity 73, member B
Gene Aliases
RGD1310887
Chromosome Location
Chr.3: 8928495 - 8951189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106566.1 NP_001100036.1 2 - 3 285 121
XM_006233781.1 XP_006233843.1 2 - 3 334 121
XM_006233782.2 XP_006233844.1 2 - 3 350 121
GenBank mRNA FQ212119.1 - 2 - 3 253 121
FQ231217.1 - 2 - 3 289 121

View Full Product Details