Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prim2
Entrez Gene ID
301323
Gene Name
primase, DNA, polypeptide 2
Gene Aliases
Pola3
Chromosome Location
Chr.9: 38535439 - 38745058 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024762.1 NP_001019933.1 11 - 12 1146 97
XM_006244686.2 XP_006244748.1 9 - 10 975 97
XM_006244689.2 XP_006244751.1 9 - 10 926 97
XM_006244690.2 XP_006244752.1 9 - 10 992 97
XM_006244691.1 XP_006244753.1 9 - 10 993 97
GenBank mRNA AJ011607.1 - 8 - 9 830 97
BC097454.1 - 11 - 12 1146 97

View Full Product Details