Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9a4
Entrez Gene ID
24785
Gene Name
solute carrier family 9, subfamily A (NHE4, cation proton antiporter 4), member 4
Gene Aliases
Nhe4
Chromosome Location
Chr.9: 47280562 - 47334752 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173098.1 NP_775121.1 6 - 7 1959 131
XM_006244738.2 XP_006244800.1 6 - 7 2125 131
XM_008766988.1 XP_008765210.1 6 - 7 2125 131
XM_008766989.1 XP_008765211.1 7 - 8 2028 131
XM_008766990.1 XP_008765212.1 7 - 8 1901 131
XM_008766991.1 XP_008765213.1 6 - 7 1248 131
GenBank mRNA M85301.1 - 6 - 7 1959 131

View Full Product Details