Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd18
Entrez Gene ID
301436
Gene Name
potassium channel tetramerization domain containing 18
Gene Aliases
RGD1564820
Chromosome Location
Chr.9: 64849547 - 64916652 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106914.1 NP_001100384.1 8 - 9 4341 84
XM_006244965.2 XP_006245027.1 6 - 7 2390 84
XM_006244966.2 XP_006245028.1 6 - 7 942 84
XM_006244967.2 XP_006245029.1 6 - 7 1454 84
XM_008767084.1 XP_008765306.1 5 - 6 3136 84
XM_008767085.1 XP_008765307.1 5 - 6 3254 84
XM_008767086.1 XP_008765308.1 6 - 7 947 84
XM_008767087.1 XP_008765309.1 6 - 7 876 84
GenBank mRNA FQ209862.1 - 7 - 8 1487 84
FQ228496.1 - 7 - 8 1510 84

View Full Product Details