The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106555.1 | NP_001100025.1 | 6 - 7 | 656 | 130 |
XM_006233581.2 | XP_006233643.1 | 6 - 7 | 1041 | 130 | |
GenBank mRNA | FQ210613.1 | - | 6 - 7 | 616 | 130 |
FQ212796.1 | - | 5 - 6 | 795 | 130 | |
FQ218819.1 | - | 6 - 7 | 616 | 130 |