Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tor1b
Entrez Gene ID
311854
Gene Name
torsin family 1, member B
Gene Aliases
-
Chromosome Location
Chr.3: 9792899 - 9798924 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039197.1 NP_001034286.1 4 - 5 784 66
GenBank mRNA BC111704.1 - 4 - 5 784 66

View Full Product Details