Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d2
Entrez Gene ID
313234
Gene Name
TBC1 domain family, member 2
Gene Aliases
-
Chromosome Location
Chr.5: 62213001 - 62261468 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107933.1 NP_001101403.1 7 - 8 1973 87
XM_006238056.2 XP_006238118.1 9 - 10 3576 87
GenBank mRNA BC168982.1 - 9 - 10 2181 87

View Full Product Details