Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tstd2
Entrez Gene ID
362514
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 2
Gene Aliases
RGD1310893
Chromosome Location
Chr.5: 61693203 - 61734011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108663.1 NP_001102133.1 2 - 3 393 133
XM_006238084.1 XP_006238146.1 2 - 3 315 133
XM_006238085.2 XP_006238147.1 2 - 3 472 133
XM_006238086.1 XP_006238148.1 1 - 2 208 133
XM_006238088.2 XP_006238150.1 2 - 3 315 133
XM_006238089.2 XP_006238151.1 2 - 3 315 133
XM_006238090.2 XP_006238152.1 2 - 3 315 133
XM_006238092.2 XP_006238154.1 2 - 3 315 133
XM_008763698.1 XP_008761920.1 2 - 3 315 133
GenBank mRNA BC158668.1 - 2 - 3 409 133

View Full Product Details