The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108663.1 | NP_001102133.1 | 9 - 10 | 1473 | 91 |
XM_006238088.2 | XP_006238150.1 | 9 - 10 | 1395 | 91 |