The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053783.1 | NP_446235.1 | 6 - 7 | 922 | 106 |
GenBank mRNA | AF201901.1 | - | 6 - 7 | 922 | 106 |
BC062039.1 | - | 6 - 7 | 864 | 106 |