Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armc10
Entrez Gene ID
296758
Gene Name
armadillo repeat containing 10
Gene Aliases
2810037c14rik, RGD1305915
Chromosome Location
Chr.4: 10004846 - 10020017 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106576.2 NP_001100046.2 2 - 3 426 86
GenBank mRNA BC079464.1 - 2 - 3 330 86
BC161913.1 - 2 - 3 431 86
DY320753.1 - 2 - 3 426 86

View Full Product Details