Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tox
Entrez Gene ID
362481
Gene Name
thymocyte selection-associated high mobility group box
Gene Aliases
-
Chromosome Location
Chr.5: 19892698 - 20189721 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108654.1 NP_001102124.1 5 - 6 902 124
XM_006237844.2 XP_006237906.1 5 - 6 1261 124
XM_008763547.1 XP_008761769.1 4 - 5 1007 124

View Full Product Details