Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kremen1
Entrez Gene ID
114107
Gene Name
kringle containing transmembrane protein 1
Gene Aliases
Kremen
Chromosome Location
Chr.14: 85441209 - 85503661 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053649.1 NP_446101.1 7 - 8 1185 97
GenBank mRNA AB065090.1 - 7 - 8 1185 97

View Full Product Details