Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dcbld2
Entrez Gene ID
155696
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
Esdn
Chromosome Location
Chr.11: 44241111 - 44292884 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130419.1 NP_569103.1 5 - 6 700 105
GenBank mRNA AF387549.1 - 5 - 6 700 105

View Full Product Details