Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcbld2
Entrez Gene ID
155696
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
Esdn
Chromosome Location
Chr.11: 44241111 - 44292884 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130419.1 NP_569103.1 7 - 8 872 83
GenBank mRNA AF387549.1 - 7 - 8 872 83

View Full Product Details