The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012753.2 | NP_036885.1 | 7 - 8 | 1269 | 76 |
XM_006231434.2 | XP_006231496.1 | 8 - 9 | 1592 | 76 | |
XM_006231435.2 | XP_006231497.1 | 8 - 9 | 1561 | 76 | |
GenBank mRNA | BC078898.1 | - | 7 - 8 | 1269 | 76 |
M21208.1 | - | 3 - 4 | 430 | 76 | |
M22204.1 | - | 7 - 8 | 1271 | 76 | |
M27282.1 | - | 3 - 4 | 424 | 76 | |
M31681.1 | - | 7 - 8 | 1281 | 76 | |
X14086.1 | - | 7 - 8 | 1271 | 76 |