The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012981.2 | NP_037113.2 | 4 - 5 | 559 | 91 |
GenBank mRNA | BC085752.1 | - | 4 - 5 | 559 | 91 |
D89863.1 | - | - | 967 | 91 |