Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp2r3a
Entrez Gene ID
363122
Gene Name
protein phosphatase 2, regulatory subunit B'', alpha
Gene Aliases
-
Chromosome Location
Chr.8: 109481352 - 109621460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012202.2 NP_001012202.2 6 - 7 2931 149
XM_006243633.2 XP_006243695.1 7 - 8 3233 149
XM_006243634.2 XP_006243696.1 6 - 7 1657 149
GenBank mRNA BC087097.1 - 6 - 7 1006 149

View Full Product Details