Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amotl2
Entrez Gene ID
65157
Gene Name
angiomotin like 2
Gene Aliases
Lccp
Chromosome Location
Chr.8: 111210252 - 111226217 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031717.1 NP_113905.1 9 - 10 2412 97
XM_006243680.2 XP_006243742.1 9 - 10 2514 97

View Full Product Details