The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009670.1 | NP_001009670.1 | 5 - 6 | 1071 | 70 |
XM_006243717.2 | XP_006243779.1 | 4 - 5 | 912 | 70 | |
XM_006243720.2 | XP_006243782.1 | 4 - 5 | 1666 | 70 | |
XM_008766494.1 | XP_008764716.1 | 5 - 6 | 1483 | 70 | |
XM_008766495.1 | XP_008764717.1 | 6 - 7 | 1137 | 70 | |
XM_008766496.1 | XP_008764718.1 | 5 - 6 | 1086 | 70 | |
XM_008766497.1 | XP_008764719.1 | 5 - 6 | 955 | 70 | |
XM_008766498.1 | XP_008764720.1 | 4 - 5 | 907 | 70 | |
GenBank mRNA | BC088472.1 | - | 5 - 6 | 1071 | 70 |