The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108175.1 | NP_001101645.1 | 13 - 14 | 2532 | 68 |
XM_006243528.2 | XP_006243590.1 | 12 - 13 | 2711 | 68 | |
XM_008766449.1 | XP_008764671.1 | 12 - 13 | 2734 | 68 |