Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zglp1
Entrez Gene ID
100360291
Gene Name
zinc finger, GATA-like protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 22081439 - 22086461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242677.2 XP_006242739.1 2 - 3 857 66

View Full Product Details