The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181626.3 | NP_853657.1 | 2 - 3 | 198 | 107 |
GenBank mRNA | AF508158.1 | - | 2 - 3 | 164 | 107 |
BC070929.1 | - | 2 - 3 | 198 | 107 | |
BC078677.1 | - | 2 - 3 | 208 | 107 |