Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Idnk
Entrez Gene ID
498695
Gene Name
idnK, gluconokinase homolog (E. coli)
Gene Aliases
-
Chromosome Location
Chr.17: 6885323 - 6892620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253564.2 XP_006253626.1 3 - 4 223 94
GenBank mRNA BC087713.1 - 3 - 4 185 94

View Full Product Details