The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017496.1 | NP_001017496.1 | 1 - 2 | 85 | 90 |
XM_006250710.2 | XP_006250772.1 | 1 - 2 | 123 | 90 | |
GenBank mRNA | BC088260.1 | - | 1 - 2 | 85 | 90 |