The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007683.1 | NP_001007684.1 | 3 - 4 | 291 | 75 |
GenBank mRNA | AY325256.1 | - | 3 - 4 | 238 | 75 |
BC079066.1 | - | 3 - 4 | 291 | 75 |