Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irf7
Entrez Gene ID
293624
Gene Name
interferon regulatory factor 7
Gene Aliases
-
Chromosome Location
Chr.1: 214249388 - 214252909 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033691.1 NP_001028863.1 4 - 5 846 130
XM_006230540.1 XP_006230602.1 5 - 6 945 130
GenBank mRNA BC104686.1 - 4 - 5 846 130
FQ232130.1 - 3 - 4 299 130
FQ232859.1 - 2 - 3 138 130

View Full Product Details