Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp11c
Entrez Gene ID
317599
Gene Name
ATPase, class VI, type 11C
Gene Aliases
Atp11c-ps1, RGD1564481
Chromosome Location
Chr.X: 143340712 - 143525588 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257596.2 XP_006257658.1 27 - 28 3150 149
XM_006257597.2 XP_006257659.1 27 - 28 4163 149
XM_006257599.2 XP_006257661.1 27 - 28 4163 149

View Full Product Details