The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012498.1 | NP_036630.1 | 8 - 9 | 863 | 70 |
GenBank mRNA | BC062034.1 | - | 8 - 9 | 848 | 70 |
X05884.1 | - | 8 - 9 | 860 | 70 |