Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcf3
Entrez Gene ID
171046
Gene Name
transcription factor 3
Gene Aliases
Pan2, Tcfe2a
Chromosome Location
Chr.7: 12146642 - 12168400 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035237.1 NP_001030314.1 2 - 3 217 138
NM_133524.2 NP_598208.2 2 - 3 217 138
GenBank mRNA AJ973227.1 - 2 - 3 217 138
BC078750.1 - 2 - 3 145 138
X54549.1 - 2 - 3 217 138
X62323.1 - 2 - 3 118 138

View Full Product Details