Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skap1
Entrez Gene ID
286975
Gene Name
src kinase associated phosphoprotein 1
Gene Aliases
Scap1, Skap55
Chromosome Location
Chr.10: 84309430 - 84619051 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173311.2 NP_775433.2 8 - 9 734 82
GenBank mRNA AB092812.1 - 8 - 9 636 82
BC097931.1 - 8 - 9 734 82

View Full Product Details