The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163320.1 | NP_001156792.1 | 8 - 9 | 1011 | 92 |
XM_006248927.2 | XP_006248989.1 | 8 - 9 | 3247 | 92 |