Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bud31
Entrez Gene ID
89819
Gene Name
BUD31 homolog (S. cerevisiae)
Gene Aliases
Edg2, G10
Chromosome Location
Chr.12: 11232963 - 11240382 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769003.1 XP_008767225.1 1 - 2 92 115

View Full Product Details