Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cnih2
Entrez Gene ID
361705
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Gene Aliases
CNIH-2, Cnih3, RGD1304930
Chromosome Location
Chr.1: 220474485 - 220480740 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025132.1 NP_001020303.1 5 - 6 709 101
XM_008760153.1 XP_008758375.1 4 - 5 1287 101
XM_008760154.1 XP_008758376.1 5 - 6 1347 101
GenBank mRNA BC091325.1 - 5 - 6 709 101

View Full Product Details