Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stx5
Entrez Gene ID
65134
Gene Name
syntaxin 5
Gene Aliases
Stx5a
Chromosome Location
Chr.1: 224939497 - 224955658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031704.2 NP_113892.2 9 - 10 918 138
XM_006230997.2 XP_006231059.1 8 - 9 1461 138
GenBank mRNA BC085724.1 - 8 - 9 1244 138
BC127489.1 - 9 - 10 918 138
L20822.1 - 8 - 9 748 138

View Full Product Details