Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Orm1
Entrez Gene ID
24614
Gene Name
orosomucoid 1
Gene Aliases
AGP, Agpa1, OMD, Orm
Chromosome Location
Chr.5: 79179668 - 79182820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053288.2 NP_445740.1 2 - 3 299 90
GenBank mRNA FQ209739.1 - 2 - 3 301 90
FQ218200.1 - 2 - 3 304 90
FQ219945.1 - 2 - 3 305 90
FQ220896.1 - 2 - 3 305 90
FQ221011.1 - 2 - 3 304 90
FQ221095.1 - 2 - 3 300 90
FQ221803.1 - 1 - 2 95 90
FQ221848.1 - 2 - 3 301 90
FQ222215.1 - 2 - 3 301 90
FQ222231.1 - 1 - 2 112 90
FQ222829.1 - 2 - 3 301 90
FQ223035.1 - 2 - 3 301 90
FQ228353.1 - 2 - 3 215 90
FQ228371.1 - 2 - 3 305 90
FQ228464.1 - 2 - 3 299 90
FQ228466.1 - 2 - 3 301 90
FQ228489.1 - 2 - 3 301 90
FQ228584.1 - 2 - 3 301 90
FQ228800.1 - 2 - 3 301 90
FQ229026.1 - 2 - 3 300 90
FQ229515.1 - 2 - 3 219 90
FQ229654.1 - 2 - 3 299 90
FQ230009.1 - 2 - 3 305 90
J00696.1 - 2 - 3 300 90
V01216.1 - 2 - 3 299 90

View Full Product Details