Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dpm3
Entrez Gene ID
502017
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 3
Gene Aliases
RGD1561807
Chromosome Location
Chr.2: 188583664 - 188584179 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109331.2 NP_001102801.1 1 - 2 51 65

View Full Product Details