Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dchs2
Entrez Gene ID
310550
Gene Name
dachsous cadherin-related 2
Gene Aliases
RGD1563329
Chromosome Location
Chr.2: 182113334 - 182319003 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232535.2 XP_006232597.1 11 - 12 5799 140

View Full Product Details