Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arl16
Entrez Gene ID
688311
Gene Name
ADP-ribosylation factor-like 16
Gene Aliases
-
Chromosome Location
Chr.10: 109636770 - 109639023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750985.2 XP_003751033.1 4 - 5 519 87

View Full Product Details