Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htra3
Entrez Gene ID
360959
Gene Name
HtrA serine peptidase 3
Gene Aliases
RGD1308120
Chromosome Location
Chr.14: 80248140 - 80276534 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271027.1 NP_001257956.1 3 - 4 893 100

View Full Product Details