Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sepsecs
Entrez Gene ID
679383
Gene Name
Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase
Gene Aliases
-
Chromosome Location
Chr.14: 60657730 - 60687950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128287.2 NP_001121759.1 4 - 5 549 88
GenBank mRNA BC166584.1 - 4 - 5 549 88

View Full Product Details