Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmp2
Entrez Gene ID
688790
Gene Name
peripheral myelin protein 2
Gene Aliases
-
Chromosome Location
Chr.2: 93827504 - 93831419 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109514.1 NP_001102984.1 3 - 4 388 116

View Full Product Details