Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sergef
Entrez Gene ID
365243
Gene Name
secretion regulating guanine nucleotide exchange factor
Gene Aliases
RGD1563497
Chromosome Location
Chr.1: 102461327 - 102664967 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229227.2 XP_006229289.1 10 - 11 1131 61
XM_006229228.2 XP_006229290.1 9 - 10 1068 61
XM_006229229.2 XP_006229291.1 8 - 9 891 61
XM_344891.7 XP_344892.4 9 - 10 1050 61

View Full Product Details