The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012782.1 | NP_036914.1 | 8 - 9 | 1172 | 77 |
GenBank mRNA | BC089915.1 | - | 6 - 7 | 819 | 77 |
BC161819.1 | - | 8 - 9 | 1177 | 77 | |
J02827.1 | - | 8 - 9 | 1156 | 77 |